Learn More
American Research Products Inc Rat L Lactate Dehydrogenase L
Catalog No. 5000329061
Encompass
Rat L-Lactate Dehydrogenase L-LDH ELISA Kit from CUSABIO Detection Range 39 mU/mL-2500 mU/mL Sensitivity 9 77 mU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29061 Supplier American Research Products Inc Supplier No. CSBE11324R
Rat L-Lactate Dehydrogenase L-LDH ELISA Kit from CUSABIO Detection Range 39 mU/mL-2500 mU/mL Sensitivity 9 77 mU/mL Sample serum plasma tissue homogenates Method Sandwich