Learn More
Biomatik Corporation Rat Lactase (LCT) ELISA Kit
Supplier:  Biomatik Corporation EKU05528
Encompass_Preferred
Rat Lactase (LCT) ELISA Kit for detecting LCT in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02401. Target Synonyms: LAC; LPH; LPH1; Lactose-Phlorizin Hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8898