Learn More
Biomatik Corporation Rat Lactate Dehydrogenase B (LDHB) ELISA Kit
Supplier:  Biomatik Corporation EKU11033
Encompass_Preferred
Rat Lactate Dehydrogenase B (LDHB) ELISA Kit for detecting LDHB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42123. Gene ID: 24534. Target Synonyms: LDH-B; LDH-H; Renal carcinoma antigen NY-REN-46. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7298