Learn More
Biomatik Corporation Rat Lactate Dehydrogenase C (LDHC) ELISA Kit
Supplier:  Biomatik Corporation EKU08471
Encompass_Preferred
Rat Lactate Dehydrogenase C (LDHC) ELISA Kit for detecting LDHC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.26ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19629. Target Synonyms: LDH-C; CT32; LDHX; LDH3; Cancer/Testis Antigen 32; LDH testis subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7753