Learn More
Biomatik Corporation Rat Laminin Alpha 1 (LAMA1) ELISA Kit
Supplier:  Biomatik Corporation EKU09009
Encompass_Preferred
Rat Laminin Alpha 1 (LAMA1) ELISA Kit for detecting LAMA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A409. Gene ID: 316758. Target Synonyms: LAM-A1; LAMA; Laminin A; LaMa1; S-laminin subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8577