Learn More
Sino Biological Anti-LAYN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80431T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Layilin / LAYN (Catalog#80431-R08H; NP_001178926.1; Met1-Glu224). Layilin / LAYN specific IgG was purified by Rat Layilin / LAYN affinity chromatography.
Catalog No. 50-240-8085