Learn More
BIOHIPPO Rat LCT Microsample
SDP
Catalog No. 5000315147
Encompass
Rat LCT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LCT in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 124 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LCT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LCT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LCT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15147 Supplier BIOHIPPO Supplier No. BHE15210895
Rat LCT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LCT in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 124 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LCT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LCT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LCT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color