Learn More
Sino Biological Anti-LDHA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80664T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat LDH-A / LDHA (rR LDH-A / LDHA; Catalog#80664-R07E; NP_058721.1; Met1-Phe332). LDH-A / LDHA specific IgG was purified by Rat LDH-A / LDHA affinity chromatography.
Catalog No. 50-255-2764