Learn More
Crystal Chem Inc Rat LDL-Cholesterol Assay Kit, 96-tests
SDP
Catalog No. 501947653
Encompass_Preferred
The LDL Cholesterol Rat Assay is used to quantify levels of LDL cholesterol in rat serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 4.5 mg/dL. Dynamic range of 4.5 - 245 mg/dL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7653 Supplier Crystal Chem Inc Supplier No. 79960
The LDL Cholesterol Rat Assay is used to quantify levels of LDL cholesterol in rat serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 4.5 mg/dL. Dynamic range of 4.5 - 245 mg/dL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.