Learn More
BIOHIPPO Rat LDLR Microsample
SDP
Catalog No. 5000315741
Encompass
Rat LDLR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LDLR in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LDLR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LDLR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LDLR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15741 Supplier BIOHIPPO Supplier No. BHE15210592
Rat LDLR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LDLR in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LDLR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LDLR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LDLR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color