Learn More
American Research Products Inc Rat Lecithin Cholesterol Acylt
Catalog No. 5000326732
Encompass
Rat Lecithin Cholesterol Acyltransferase (LCAT) ELISA Kit from CUSABIO Detection Range 1 87 ng/mL-120 ng/mL Sensitivity 0 94 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26732 Supplier American Research Products Inc Supplier No. CSBE14201R
Rat Lecithin Cholesterol Acyltransferase (LCAT) ELISA Kit from CUSABIO Detection Range 1 87 ng/mL-120 ng/mL Sensitivity 0 94 ng/mL Sample serum plasma tissue homogenates Method Competitive