Learn More
Biomatik Corporation Rat Left/Right Determination Factor 1 (LEFTY1) CLIA Kit
Supplier:  Biomatik Corporation EKU09830
Encompass_Preferred
Rat Left/Right Determination Factor 1 (LEFTY1) CLIA Kit for detecting LEFTY1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: D4A670. Gene ID: 498299. Target Synonyms: LEFTY-B; LEFTYB; LEFT-B; LEFTB; Left Right Determination Factor B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8424