Learn More
Crystal Chem Inc Rat Leptin ELISA KIT, 96-wells, 5UL
SDP
Supplier:  Crystal Chem Inc 90040
Encompass
The Rat Leptin ELISA is used to quantify levels of rat leptin in serum, plasma, cell culture, and fluid using only a 5 µL sample volume. Highlights: Kits use a sample volume of only 5 µL. High Sensitivity of 200 pg/mL. Dynamic Range of 0.2 - 12.8 ng/mL. Designed for rat samples. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7915