Learn More
Bioworld RAT LEPTIN (RECOMBINANT)
Supplier:  Bioworld 220606701
Encompass_Preferred
Recombinant Rat Leptin (Anti Obesity Protein) Source: E.Coli Background: Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe heredit...
Catalog No. 50-253-766