Learn More
Biomatik Corporation Rat Leucine Aminopeptidase (LAP) ELISA Kit
Supplier:  Biomatik Corporation EKU05602
Encompass_Preferred
Rat Leucine Aminopeptidase (LAP) ELISA Kit for detecting LAP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q68FS4. Gene ID: 289668. Target Synonyms: LAPEP, LAP3; PEPS; Peptidase S; Leucyl Aminopeptidase; Cytosol aminopeptidase; Proline aminopeptidase; Prolyl aminopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7242