Learn More
BIOHIPPO Rat LFA1a Microsample
SDP
Catalog No. 5000315373
Encompass
Rat LFA1a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LFA1a in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LFA1a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LFA1a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LFA1a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15373 Supplier BIOHIPPO Supplier No. BHE15210971
Add to Cart
Add to Cart
Rat LFA1a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LFA1a in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LFA1a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LFA1a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LFA1a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color