Learn More
Sino Biological Anti-Galectin-1/LGALS1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80248RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Galectin-1 (rR Galectin-1 ; Catalog#80248-RNAE; P11762; Ala2-Glu135). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2797