Learn More
BIOHIPPO Rat LGALS3BP Microsample
SDP
Catalog No. 5000315496
Encompass
Rat LGALS3BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LGALS3BP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 68 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LGALS3BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LGALS3BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LGALS3BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15496 Supplier BIOHIPPO Supplier No. BHE15210800
Rat LGALS3BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LGALS3BP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 68 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LGALS3BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LGALS3BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LGALS3BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color