Learn More
Biomatik Corporation Rat Lipin 1 (LPIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU05655
Encompass_Preferred
Rat Lipin 1 (LPIN1) ELISA Kit for detecting LPIN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5XIM8. Gene ID: 313977. Target Synonyms: Phosphatidate phosphatase LPIN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7315