Learn More
Biomatik Corporation Rat Lipocalin 1 (LCN1) ELISA Kit
Supplier:  Biomatik Corporation EKU11044
Encompass_Preferred
Rat Lipocalin 1 (LCN1) ELISA Kit for detecting LCN1 in serum, plasma, saliva, tears, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.246ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20289. Gene ID: 65039. Target Synonyms: TP; PMFA; VEGP; Tlc; Tear Prealbumin; Tear lipocalin; Protein Migrating Faster Than Albumin; Von Ebner Gland Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8240