Learn More
American Research Products Inc Rat lipolysaccharide binding p
Catalog No. 5000330111
Encompass
Rat lipolysaccharide binding protein LBP ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-30111 Supplier American Research Products Inc Supplier No. CSBE11184R
Rat lipolysaccharide binding protein LBP ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich