Learn More
Biomatik Corporation Rat Lipopolysaccharide Binding Protein (LBP) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10295
Encompass_Preferred
Rat Lipopolysaccharide Binding Protein (LBP) High-sensitive ELISA Kit for detecting LBP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63313. Target Synonyms: LPS-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8351