Learn More
American Research Products Inc Rat Lipopolysaccharides LPS
Catalog No. 5000326733
Encompass
Rat Lipopolysaccharides (LPS) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26733 Supplier American Research Products Inc Supplier No. CSBE14247R
Rat Lipopolysaccharides (LPS) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich