Learn More
American Research Products Inc Rat lipoprotein associated pho
Catalog No. 5000329592
Encompass
Rat lipoprotein-associated phospholipase A2 Lp-PLA2 ELISA Kit from CUSABIO Detection Range 0 78 IU/mL-50 IU/mL Sensitivity 0 195 IU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29592 Supplier American Research Products Inc Supplier No. CSBE08320R
Rat lipoprotein-associated phospholipase A2 Lp-PLA2 ELISA Kit from CUSABIO Detection Range 0 78 IU/mL-50 IU/mL Sensitivity 0 195 IU/mL Sample serum plasma Method Sandwich