Learn More
Zyagen Labs Rat Liver Genomic DNA, 0.1mg
SDP
Catalog No. NC2146510
Encompass_Preferred
Rat Liver genomic DNA is a highly pure intact high molecular size DNA. It is extracted from single tissue of single donor and treated with DNase-free RNase to remove contaminant RNA. Genomic DNA is precisely measured by Nanodrop (a spectrophotometer technology) and stored at -80oCApplications:PCR Amplification, Southern blot analysis, genomic DNA methylation, single-nucleotide polymorphism (SNP) Analysis, profiling studies in gene expression, and DNA library construction
This item is not returnable. View return policy
Catalog No. NC2146510 Supplier Zyagen Labs Supplier No. RG314
Rat Liver genomic DNA is a highly pure intact high molecular size DNA. It is extracted from single tissue of single donor and treated with DNase-free RNase to remove contaminant RNA. Genomic DNA is precisely measured by Nanodrop (a spectrophotometer technology) and stored at -80oCApplications:PCR Amplification, Southern blot analysis, genomic DNA methylation, single-nucleotide polymorphism (SNP) Analysis, profiling studies in gene expression, and DNA library construction