Learn More
Biomatik Corporation Rat Liver Receptor Homolog 1 (LRH1) ELISA Kit
Supplier:  Biomatik Corporation EKU05669
Encompass_Preferred
Rat Liver Receptor Homolog 1 (LRH1) ELISA Kit for detecting LRH1 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QWM1. Gene ID: 60349. Target Synonyms: NR5A2; B1F; B1F2; CPF; FTF; FTZ-F1; LRH-1; hB1F; hB1F-2; Hepatocytic transcription factor; Alpha-1-fetoprotein transcription factor; Nuclear Receptor Subfamily 5,Group A,Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6144