Learn More
Cayman Chemical Rat Liver X Receptor Alpha Reporter Assay System, 96 Well
SDP
Catalog No. 502909214
Encompass_Preferred
rLXRα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00311, R00311-32]
Catalog No. 50-290-9214 Supplier Cayman Chemical Supplier No. 3615396 WELL
rLXRα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R00311, R00311-32]