Learn More
Cayman Chemical Rat LIvR X RECPTR Beta Rep
SDP
Catalog No. 502910048
Encompass_Preferred
This Rat Liver X Receptor Beta (rLXRβ) assay kit is an all-inclusive firefly luciferase reporter assay system that includes, in addition to rLXRβ Reporter Cells, two optimized media for use during cell culture and (optionally) in diluting the test samples, a reference agonist, Luciferase Detection Reagent, a cell culture-ready assay plate, and a detailed protocol.
Catalog No. 50-291-0048 Supplier Cayman Chemical Supplier No. 367133X32 WELL
This Rat Liver X Receptor Beta (rLXRβ) assay kit is an all-inclusive firefly luciferase reporter assay system that includes, in addition to rLXRβ Reporter Cells, two optimized media for use during cell culture and (optionally) in diluting the test samples, a reference agonist, Luciferase Detection Reagent, a cell culture-ready assay plate, and a detailed protocol.