Learn More
American Research Products Inc Rat Low Density Lipoprotein ch
Catalog No. 5000326660
Encompass
Rat Low Density Lipoprotein cholesterol (LDL-C) ELISA kit from CUSABIO Detection Range 125 ng/mL-8000 ng/mL Sensitivity 62 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26660 Supplier American Research Products Inc Supplier No. CSBE16561R
Rat Low Density Lipoprotein cholesterol (LDL-C) ELISA kit from CUSABIO Detection Range 125 ng/mL-8000 ng/mL Sensitivity 62 5 ng/mL Sample serum plasma tissue homogenates Method Competitive