Learn More
American Research Products Inc Rat low density lipoprotein L
Catalog No. 5000329039
Encompass
Rat low density lipoprotein LDL ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 156 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29039 Supplier American Research Products Inc Supplier No. CSBE11704R
Rat low density lipoprotein LDL ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 156 g/mL Sample serum plasma tissue homogenates Method Competitive