Learn More
American Research Products Inc Rat low density lipoprotein re
Catalog No. 5000329252
Encompass
Rat low density lipoprotein receptor LDLR ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29252 Supplier American Research Products Inc Supplier No. CSBE13545R
Rat low density lipoprotein receptor LDLR ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich