Learn More
BIOHIPPO Rat LPO Microsample
SDP
Catalog No. 5000315369
Encompass
Rat LPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LPO in serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 076 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15369 Supplier BIOHIPPO Supplier No. BHE15210614
Rat LPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LPO in serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 076 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color