Learn More
Biomatik Corporation Rat Luteinizing Hormone (LH) CLIA Kit
Supplier:  Biomatik Corporation EKU10050
Encompass_Preferred
Rat Luteinizing Hormone (LH) CLIA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-8,000pg/ml. Sensitivity: 11.73pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P16235. Gene ID: 25477. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6101