Learn More
American Research Products Inc Rat luteinizing hormone LH E
Catalog No. 5000329029
Encompass
Rat luteinizing hormone (LH) Elisa kit from CUSABIO Detection Range 0 3 mlU/ml-60 mlU/ml Sensitivity 0 15 mlU/ml Sample serum plasma Method Competitive
Catalog No. 50-003-29029 Supplier American Research Products Inc Supplier No. CSBE12654R
Rat luteinizing hormone (LH) Elisa kit from CUSABIO Detection Range 0 3 mlU/ml-60 mlU/ml Sensitivity 0 15 mlU/ml Sample serum plasma Method Competitive