Learn More
Biomatik Corporation Rat Lymphocyte Function Associated Antigen 1 Alpha (CD11a) ELISA Kit
Supplier:  Biomatik Corporation EKU05704
Encompass_Preferred
Rat Lymphocyte Function Associated Antigen 1 Alpha (CD11a) ELISA Kit for detecting CD11a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3T1L6. Gene ID: 308995. Target Synonyms: LFA1a; ITG-AL; LFA-1; LFA1-A; ITGAL; Integrin,Alpha L; CD11 antigen-like family member A; Leukocyte function-associated molecule 1 alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5767