Learn More
American Research Products Inc Rat Lysozyme C LYZ ELISA kit
Catalog No. 5000327449
Encompass
Rat Lysozyme C (LYZ) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27449 Supplier American Research Products Inc Supplier No. CSBEL013283RA
Rat Lysozyme C (LYZ) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Competitive