Learn More
Biomatik Corporation Rat Lysyl Oxidase (LOX) ELISA Kit
Supplier:  Biomatik Corporation EKU09128
Encompass_Preferred
Rat Lysyl Oxidase (LOX) ELISA Kit for detecting LOX in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.293ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16636. Gene ID: 24914. Target Synonyms: Protein-Lysine 6-Oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7390