Learn More
American Research Products Inc Rat Macrophage Colony Stimulat
Catalog No. 5000326667
Encompass
Rat Macrophage Colony-Stimulating Factor M-CSF ELISA kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26667 Supplier American Research Products Inc Supplier No. CSBE07424R
Rat Macrophage Colony-Stimulating Factor M-CSF ELISA kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich