Learn More
Biomatik Corporation Rat Macrophage Inflammatory Protein 3 Alpha (MIP3a) ELISA Kit
Supplier:  Biomatik Corporation EKU05754
Encompass_Preferred
Rat Macrophage Inflammatory Protein 3 Alpha (MIP3a) ELISA Kit for detecting MIP3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97884. Gene ID: 29538. Target Synonyms: CCL20; CKb4; LARC; MIP3-A; SCYA20; ST38; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 20; Beta-Chemokine Exodus-1; Liver And Activation-Regulated Chemokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5032