Learn More
American Research Products Inc Rat Macrophage Migration Inhib
Catalog No. 5000328974
Encompass
Rat Macrophage Migration Inhibitory Factor MIF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28974 Supplier American Research Products Inc Supplier No. CSBE07293R
Rat Macrophage Migration Inhibitory Factor MIF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich