Learn More
Biomatik Corporation Rat Macrophage Migration Inhibitory Factor (MIF) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10562
Encompass_Preferred
Rat Macrophage Migration Inhibitory Factor (MIF) Wide-range ELISA Kit for detecting MIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30904. Gene ID: 81683. Target Synonyms: GIF; GLIF; MMIF; Glycosylation-Inhibiting Factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6299