Learn More
BIOHIPPO Rat MAG Microsample
SDP
Catalog No. 5000315582
Encompass
Rat MAG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAG in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 131 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MAG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MAG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MAG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15582 Supplier BIOHIPPO Supplier No. BHE15211768
Rat MAG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAG in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 131 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MAG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MAG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MAG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color