Learn More
Biomatik Corporation Rat Malate Dehydrogenase 1 (MDH1) ELISA Kit
Supplier:  Biomatik Corporation EKU05780
Encompass_Preferred
Rat Malate Dehydrogenase 1 (MDH1) ELISA Kit for detecting MDH1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88989. Gene ID: 24551. Target Synonyms: MDHA; Malate Dehydrogenase 1,NAD (Soluble) ; Cytosolic malate dehydrogenase; Diiodophenylpyruvate reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8885