Learn More
BIOHIPPO Rat MAOB Microsample
SDP
Catalog No. 5000314115
Encompass
Rat MAOB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAOB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 33 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MAOB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MAOB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MAOB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14115 Supplier BIOHIPPO Supplier No. BHE15210702
Rat MAOB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MAOB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 33 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MAOB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MAOB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MAOB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color