Learn More
American Research Products Inc Rat matriX metallopeptidase 8
Catalog No. 5000326444
Encompass
Rat matriX metallopeptidase 8 (neutrophil collagenase) (MMP8) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26444 Supplier American Research Products Inc Supplier No. CSBE07406R
Rat matriX metallopeptidase 8 (neutrophil collagenase) (MMP8) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich