Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 13 (MMP13) CLIA Kit
Supplier:  Biomatik Corporation EKU09577
Encompass_Preferred
Rat Matrix Metalloproteinase 13 (MMP13) CLIA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P23097. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5179