Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 2 (MMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09580
Encompass_Preferred
Rat Matrix Metalloproteinase 2 (MMP2) CLIA Kit for detecting MMP2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P33436. Gene ID: 81686. Target Synonyms: CLG4; CLG4A; MMP-II; MONA; TBE-1; PEX; Gelatinase A; 72kDa Gelatinase; 72kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7438