Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 8 (MMP8) CLIA Kit
Supplier:  Biomatik Corporation EKU09449
Encompass_Preferred
Rat Matrix Metalloproteinase 8 (MMP8) CLIA Kit for detecting MMP8 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.48pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O88766. Gene ID: 63849. Target Synonyms: CLG2; HNC; PMNL-CL; Collagenase 2; Neutrophil Collagenase; PMNL collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5640