Learn More
BIOHIPPO Rat MC4R Microsample
SDP
Catalog No. 5000315365
Encompass
Rat MC4R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MC4R in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MC4R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MC4R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MC4R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15365 Supplier BIOHIPPO Supplier No. BHE15210594
Rat MC4R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MC4R in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MC4R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MC4R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MC4R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color