Learn More
Biomatik Corporation Rat Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit
Supplier:  Biomatik Corporation EKU05898
Encompass_Preferred
Rat Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit for detecting MIA1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62946. Gene ID: 81510. Target Synonyms: CD-RAP; Melanoma-derived growth regulatory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8876